Learn More
Novus Biologicals™ PGK1 Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP130187
Catalog No. NBP130187
Add to cart
Description
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-417 of Human PGK1 The Recombinant Human PGK1 Protein is derived from E. coli. The Recombinant Human PGK1 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
NP_000282
20mM Tris buffer (pH 8.0), 10% glycerol, 1mM DTT
46.8kDa
Protein
MGSSHHHHHH SSGLVPRGSH MSLSNKLTLD KLDVKGKRVV MRVDFNVPMK NNQITNNQRI KAAVPSIKFC LDNGAKSVVL MSHLGRPDGV PMPDKYSLEP VAVELKSLLG KDVLFLKDCV GPEVEKACAN PAAGSVILLE NLRFHVEEEG KGKDASGNKV KAEPAKIEAF RASLSKLGDV YVNDAFGTAH RAHSSMVGVN LPQKAGGFLM KKELNYFAKA LESPERPFLA ILGGAKVADK IQLINNMLDK VNEMIIGGGM AFTFLKVLNN MEIGTSLFDE EGAKIVKDLM SKAEKNGVKI TLPVDFVTAD KFDENAKTGQ ATVASGIPAG WMGLDCGPES SKKYAEAVTR AKQIVWNGPV GVFEWEAFAR GTKALMDEVV KATSRGCITI IGGGDTATCC AKWNTEDKVS HVSTGGGASL ELLEGKVLPG VDALSNI
ELISA, SDS-Page
5230
PGK1 Protein
>95%
−80°C. Avoid freeze-thaw cycles.