Learn More
Novus Biologicals™ PiColorLock™ Gold Colorimetric Assay Kit
Colorimetric Assay Kit
$286.35 - $631.35
Specifications
Product Type PiColorLock Gold Colorimetric Assay Kit
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
30-301-25
View Documents
Novus Biologicals™
3030125
2500 Assays
Each for $631.35
Add to cart
30-300-30
View Documents
Novus Biologicals™
3030030
625 Assays
Each for $286.35
Add to cart
Description
Description
PiColorLock phosphate detection reagent changes color in the presence of inorganic phosphate (Pi). This can be exploited to measure any enzyme that generates Pi including ATPases, GTPases, phosphatases, heat-shock proteins and DNA unwinding proteins.
Specifications
Specifications
PiColorLock Gold Colorimetric Assay Kit