Learn More
Novus Biologicals™ PiColorLock™ Gold Colorimetric Assay Kit
Colorimetric Assay Kit
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 3030030
 View more versions of this product
Catalog No. 30-300-30
  Check Availability
Add to cart
Specifications
Description
PiColorLock phosphate detection reagent changes color in the presence of inorganic phosphate (Pi). This can be exploited to measure any enzyme that generates Pi including ATPases, GTPases, phosphatases, heat-shock proteins and DNA unwinding proteins.
Order Info
Quantity
Description & Specifications
Specifications
625 Assays
Store at 4°C. Do not freeze.
PiColorLock Gold Colorimetric Assay Kit