Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ PITPN Protein
Highly purified. Generating reliable and reproducible results.
$398.00 - $1110.00
Specifications
Concentration | 1mg/mL |
---|---|
For Use With (Application) | SDS-PAGE |
Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 1mM EDTA |
Molecular Weight (g/mol) | 33.9kDa |
Purification Method | SDS-PAGE |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15091001
|
Novus Biologicals™
NBP1509100.1MG |
0.1mg |
Each for $398.00
|
|
|||||
NBP1509105
|
Novus Biologicals™
NBP1509100.5MG |
0.5mg |
Each for $1,110.00
|
|
|||||
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-270 of Human PITPN The Recombinant Human PITPN Protein is derived from E. coli. The Recombinant Human PITPN Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 1mM EDTA | |
SDS-PAGE | |
RUO | |
Unconjugated |
SDS-PAGE | |
33.9kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
PITPN | |
>95% |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title