Learn More
Novus Biologicals™ PKR Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176730PEP
Catalog No. NBP176730E
Add to cart
Description
Description
A blocking peptide containing 14 amino acids near the amino terminus of human PKR. Source: Synthetic (Accession #: AAP57628) The PKR Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
5610
For blocking activity of PKR, NBP1-76730
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
14 amino acids near the amino terminus of human PKR
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
PKR