Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Polyinosinic-polycytidylic acid

Applications: Functional, In vitro assay, In vivo assay, Ligand Activation

$277.00 - $421.50

Specifications

For Use With (Application) Used for Activation of TLR3. Stimulation of TLR3 Can be Achieved with 10-100 µg/mL Use in In Vitro Assay Reported in Scientific Literature (PMID 24743304) .Use in Ligand Activation, and Functional Reported in Scientific Literature (PMID 25957979). Use in In Vivo Assay Reported in Scientific Literature (PMID 26196739). Use in Ligand Activation Reported in Scientific Literature (PMID 26503951)
Storage Requirements Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Product Type Polyinosinic-polycytidylic Acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP225288
View Documents
Novus Biologicals™
NBP225288
10 mg
Each for $277.00
Add to cart
 
NBP22528850
View Documents
Novus Biologicals™
NBP22528850MG
50mg
Each for $421.50
Add to cart
 
Description

Description

Polyinosinic-polycytidylic acid (Poly(I:C)) is a synthetic analog of double stranded RNA (dsRNA) that is produced during the replicative life cycle of a majority of viruses. Poly(I:C) is known to induce type-1 interferon (IFN) through TLR3. Poly(I:C) directly binds to TLR3.

  • Species: Human, Mouse
  • Stimulation of TLR3 can be achieved with 10-100μg/mL
Specifications

Specifications

Used for Activation of TLR3. Stimulation of TLR3 Can be Achieved with 10-100 µg/mL Use in In Vitro Assay Reported in Scientific Literature (PMID 24743304) .Use in Ligand Activation, and Functional Reported in Scientific Literature (PMID 25957979). Use in In Vivo Assay Reported in Scientific Literature (PMID 26196739). Use in Ligand Activation Reported in Scientific Literature (PMID 26503951)
Store at 4 C short term. Aliquot and store (at -20 C long term. Avoid freeze-thaw cycles.)
Polyinosinic-polycytidylic Acid
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit