Learn More
Novus Biologicals™ PPP2R1B Overexpression Lysate (Native)
Manufacturer:  Novus Biologicals™ NBP208078
Catalog No. NBP208078
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The PPP2R1B Overexpression Lysate is derived from human.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_277. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens PPP2R1B, mRNA.
Native
0.1mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot