Learn More
Novus Biologicals™ PTH2R Overexpression Lysate (Native)
Supplier: Novus Biologicals™ NBL114947
Description
The PTH2R Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC27775 and based on accession number NM_548. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens parathyroid hormone 2 receptor (PTH2R), mRNA. | |
Native |