Learn More
Novus Biologicals™ Rabbit Rabbit IgG, Fab2 Protein
Rabbit Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197017
Catalog No. NBP197017
Add to cart
Description
Description
IgG F(ab)2 Isotype Control antibody specifically detects IgG F(ab)2 in Rabbit samples. It is validated for Western Blot, ELISA, Immunoprecipitation, SDS-Page.
Specifications
Specifications
WB, ELISA, IP
Rabbit IgG, Fab2 Protein
Rabbit
Rabbit
Potassium Phosphate, 0.15M Sodium Chloride pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG, anti-Rabbit IgG F(ab)2 and anti-Rabbit Serum. No reaction was observed against anti-Rabbit IgG F(c) or anti-Pepsin.