Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Novus Biologicals™ RAIDD/CRADD Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177061PEP
Catalog No. NBP177061E
Description
Description
A blocking peptide containing 19 amino acids near the center of human RAIDD. Source: Synthetic (Accession #: AAB42217) The RAIDD/CRADD Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
8738
For Blocking the binding of NBP1-77061
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
19 amino acids near the center of human RAIDD
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
RAIDD/CRADD