Learn More
Novus Biologicals™ RAIDD/CRADD Overexpression Lysate
Supplier: Novus Biologicals™ NBL109459
Description
The RAIDD/CRADD Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
The lysate was created in HEK293T cells, using plasmid ID RC225 and based on accession number NM_385. The protein contains a C-terminal DDK tag. | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
Western Blot | |
RIPA buffer | |
Homo sapiens CASP2 and RIPK1 domain containing adaptor with death domain (CRADD), mRNA. | |
Native |