Learn More
Novus Biologicals™ Rat Rat IgG, Fc Protein
Rat Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP196967
Catalog No. NBP196967
Add to cart
Description
Description
IgG Fc Isotype Control antibody specifically detects IgG Fc in Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page.
Specifications
Specifications
ELISA, IP
Rat IgG, Fc Protein
Rat
Rat
Potassium Phosphate, 0.15M Sodium Chloride pH 7.2.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Rat IgG F(c) fragment was assayed by immunoelectrophoresis resulted in a single precipitin arc against anti-Rat Serum, anti-Rat IgG and anti-Rat IgG F(c). No reaction was observed against anti-Rat IgG F(ab)2 or anti-Papain.