Learn More
Novus Biologicals™ RCC4 Whole Cell Lysate
Manufacturer:  Novus Biologicals™ NBP130412
Catalog No. NBP130412
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
The RCC4 Whole Cell Lysate is derived from human. It has been validated for the following applications: Western Blot, SDS-Page.
Specifications
Specifications
Digestive/Waste
Standard RIPA buffer including protease and phosphatase inhibitors and 0.1% SDS diluted in 1X lithium dodecyl sulfate (LDS) sample buffer with bromophenol blue and 40-70% glycerol, pH 8.4. BME has not been added.
-80°C. Avoid freeze-thaw cycles.
Denatured
1mg
Protein
Whole
SDS-Page, Western Blot