Learn More
Novus Biologicals™ REEP2 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP177104PEP
Description
A blocking peptide containing 15 amino acids near the center of human REEP2. Source: Synthetic (Accession #: EAW62139) The REEP2 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
51308 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-77104. For Further Blocking Peptide Related Protocol, Click Here . | |
REEP2 | |
0.05 mg | |
Synthetic |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
C5orf19receptor expression-enhancing protein 2, chromosome 5 open reading frame 19, receptor accessory protein 2, SGC32445receptor expression enhancing protein 2 | |
REEP2 | |
RUO - research use only | |
15 amino acids near the center of human REEP2 |