Learn More
Novus Biologicals™ S100A13 Overexpression Lysate (Native)
Manufacturer:  Novus Biologicals™ NBP208655
Catalog No. NBP208655
Add to cart
Description
Description
The S100A13 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_119381. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens S100A13, mRNA.
Native
0.1mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot