Learn More
Novus Biologicals™ SCUBE2 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP177186PEP
Description
A blocking peptide containing 17 amino acids near the center of human SCUBE2. Source: Synthetic (Accession #: NP_066025) The SCUBE2 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
57758 | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-77186. For Further Blocking Peptide Related Protocol, Click Here . | |
SCUBE2 | |
0.05 mg | |
Synthetic |
Human | |
PBS pH 7.2 (10 mM NaH2PO4, 10 mM Na2HPO4, 130 mM NaCl) containing 0.1% bovine serum albumin. | |
Cegb1, Cegf1, CEGP1, FLJ16792, FLJ35234, MGC133057, Protein CEGP1, signal peptide, CUB and EGF-like domain-containing protein 2, signal peptide, CUB domain, EGF-like 2 | |
SCUBE2 | |
RUO - research use only | |
17 amino acids near the center of human SCUBE2 |