Learn More
Novus Biologicals™ Sheep IgG, Fab Protein
Sheep Isotype Control Antibody
Manufacturer:  Novus Biologicals™ NBP197012
Catalog No. NBP197012
Add to cart
Description
Description
IgG F(ab) Isotype Control antibody specifically detects IgG F(ab) in Sheep samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
Specifications
WB, ELISA, IP
Sheep IgG, Fab Protein
Sheep
Sheep
0.01 M Sodium Phosphate, 0.14M Sodium Chloride, pH 7.4.
4°C short term, −20°C long term. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Sheep Serum, anti-Sheep IgG and anti-Sheep IgG F(ab)2. No reaction was observed against anti-Sheep IgG F(c) or anti- Papain.