Learn More
Novus Biologicals™ SLC22A10 Antibody Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Supplier: Novus Biologicals™ NBP191568PEP
Description
A blocking peptide from human SLC22A10. Source: Synthetic Amino Acid Sequence: (Accession #: NP_001034841) QTLRVALACLGIGCSAATFSSVAVHFIELIPTVLRARASGIDLTASRIGA The SLC22A10 Blocking Peptide is derived from Synthetic. The SLC22A10 Blocking Peptide has been validated for the following applications: Antibody Competition.Specifications
387775 | |
Q63ZE4 | |
Lyophilized with sterile distilled water. | |
member 10, solute carrier family 22, member 10 | |
60kDa | |
100 μg |
Human | |
Store at -20 C. Avoid freeze-thaw cycles. | |
This Peptide is Useful as a Blocking Peptide for NBP1-91568. This Synthetic Peptide is Designed for Use in an Antibody Competition Assay with Its Corresponding Antibody. Use of This Product in Any Other Assay Has Not Yet Been Tested. For Further Blocking Peptide Related Protocol, Click Here . | |
SLC22A10 | |
SLC22A10 | |
RUO - research use only |