Learn More
Novus Biologicals™ SLC35C2 Overexpression Lysate
Supplier: Novus Biologicals™ NBP205148
Description
The SLC35C2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_77596. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens SLC35C2, mRNA. | |
Native |