Learn More
Novus Biologicals™ SnoN Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177306PEP
Catalog No. NBP177306E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the amino terminus of human SnoN. Source: Synthetic (Accession #: NP_005405) The SnoN Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
6498
For blocking activity of SnoN, NBP1-77306
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the amino terminus of human SnoN
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
SnoN