Learn More
Novus Biologicals™ Staufen Overexpression Lysate (Native)
Manufacturer:  Novus Biologicals™ NBP206974
Catalog No. NBP206974
Add to cart
Description
Description
The Staufen Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
The lysate was created in HEK293T cells, using plasmid ID RC215316 and based on accession number NP_59346. The protein contains a C-terminal DDK tag.
RIPA buffer
Homo sapiens Staufen, mRNA.
Native
0.1mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot