Learn More
Novus Biologicals™ STEAP2 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176823PEP
Catalog No. NBP176823E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the carboxy terminus of human STEAP2. Source: Synthetic (Accession #: Q8NFT2) The STEAP2 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
261729
For Blocking the binding of NBP1-76823
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the carboxy terminus of human STEAP2
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
STEAP2