Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ SurA Protein

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

$320.85 - $895.85

Specifications

Common Name SurA
Conjugate Unlabeled
For Use With (Application) ELISA, Functional Assay, SDS-Page
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1460345
View Documents
Novus Biologicals™
NBP1460340.5MG
0.5mg Each for $895.85
Add to cart
 
NBP14603401
View Documents
Novus Biologicals™
NBP1460340.1MG
0.1mg Each for $320.85
Add to cart
 
Description

Description

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 21-428 of E.coli SurA The Recombinant E. coli SurA Protein is derived from E. coli. The Recombinant E. coli SurA Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
Specifications

Specifications

SurA
ELISA, Functional Assay, SDS-Page
RUO
Unlabeled
Store at -80°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit