Learn More
Novus Biologicals™ TCF-3/E2A Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177119PEP
Catalog No. NBP177119E
Add to cart
Description
Description
A blocking peptide containing 17 amino acids near the amino terminus of human TCF3. Source: Synthetic (Accession #: NP_003191) The TCF-3/E2A Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
6929
For Blocking the binding of NBP1-77119
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
17 amino acids near the amino terminus of human TCF3
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
TCF-3/E2A