Learn More
Novus Biologicals™ TEM7/PLXDC1 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176833PEP
Catalog No. NBP176833E
Add to cart
Description
Description
A blocking peptide containing 16 amino acids near the center of human TEM7. Source: Synthetic (Accession #: AAG00869) The TEM7/PLXDC1 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
57125
For Blocking the binding of NBP1-76833
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
16 amino acids near the center of human TEM7
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
TEM7/PLXDC1