Learn More
Novus Biologicals™ THEM4 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177360PEP
Catalog No. NBP177360E
Add to cart
Description
Description
A blocking peptide containing 18 amino acids near the center of human THEM4. Source: Synthetic (Accession #: NP_444283) The THEM4 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
117145
For Blocking the binding of NBP1-77360
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
18 amino acids near the center of human THEM4
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
THEM4