Learn More
Novus Biologicals™ TLR3 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP176660PEP
Catalog No. NBP176660E
Add to cart
Description
Description
A blocking peptide containing 15 amino acids near the amino terminus of human TLR3. Source: Synthetic (Accession #: O15455) The TLR3 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
7098
For Blocking the binding of NBP1-76660
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
15 amino acids near the amino terminus of human TLR3
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
TLR3