Learn More
Novus Biologicals™ TLR7 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177205PEP
Catalog No. NBP177205E
Add to cart
Description
Description
A blocking peptide from 14 amino acids from the middle of mouse TLR7. Source: Synthetic (Accession #: NP_573474) The TLR7 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
51284
For Blocking the binding of NBP1-77205
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
14 amino acids from the middle of mouse TLR7
0.05mg
Synthetic
Mouse
−20°C. Avoid freeze-thaw cycles.
TLR7