Learn More
Novus Biologicals™ TMEM97 Peptide
Manufacturer:  Novus Biologicals™ NBP130437PEP
Catalog No. NBP130437E
Add to cart
Description
Description
A peptide to TMEM97.
Specifications
Specifications
27346
Blocking peptide for NBP1-30437 only.
RUO
TMEM97
0.05mL
Peptide dissolved in dH2O. Contains no BSA.
−80°C. Avoid freeze-thaw cycles.