Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ TNF-alpha Protein

Manufacturer:  Novus Biologicals™ NBP1428990.1MG

 View more versions of this product

Catalog No. NBP1428991M

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ELISA, Immunoassay
0.1mg
TNF-alpha
RUO
Unlabeled
Sterile liquid; 10mM sodium phosphate, pH 7.2, 150mM NaCl, with no carriers or preservatives. 0.22μm filtered.
Mouse
Store at -80°C. Avoid freeze-thaw cycles.
The recombinant mouse TNF-alpha; has been tested by sandwich ELISA using the mouse TNF-alpha; ELISA antibody pair (TN3-19.12/polyclonal antibody).
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit