Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ TPK1 Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP199102100UG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-243 of Human TPK1 The Recombinant Human TPK1 Protein is derived from E. coli. The Recombinant Human TPK1 Protein has been validated for the following applications: SDS-Page.Specifications
1mg/mL | |
Liquid. 20mM Tris-HCl buffer (pH 8.0) containing 20% glycerol, 1mM DTT | |
SDS-PAGE | |
100μg | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
TPK1 |
SDS-Page | |
29.8kDa | |
>90% | |
Human | |
RUO | |
Unlabeled |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title