Learn More
Novus Biologicals™ TRAILR3/TNFRSF10C Blocking Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177295PEP
Catalog No. NBP177295E
Add to cart
Description
Description
A blocking peptide containing 13 amino acids near the extracellular domain of human TRAIL R3 . Source: Synthetic (Accession #: AF012536) The TRAILR3/TNFRSF10C Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
8794
For blocking the activity of DcR1, NBP1-77295
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
13 amino acids near the extracellular domain of human DcR1
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
TRAIL R3/TNFRSF10C