Learn More
Novus Biologicals™ TRAIL/TNFSF10 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177062PEP
Catalog No. NBP177062E
Add to cart
Description
Description
A blocking peptide containing 17 amino acids near the carboxy terminus of human TRAIL. Source: Synthetic (Accession #: NP_003801) The TRAIL/TNFSF10 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
8743
For Blocking the binding of NBP1-77062
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
RUO
17 amino acids near the carboxy terminus of human TRAIL
0.05mg
Synthetic
Human
−20°C. Avoid freeze-thaw cycles.
TRAIL/TNFSF10