Learn More
Novus Biologicals™ TRPC6 Peptide
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP177261PEP
Catalog No. NBP177261E
Add to cart
Description
Description
A blocking peptide containing 14 amino acids near the amino terminus of human TRPC6. Source: Synthetic (Accession #: AAH93658) The TRPC6 Blocking Peptide has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Human
0.05mg
Synthetic
RUO
14 amino acids near the amino terminus of human TRPC6
7225
For Blocking the binding of NBP1-77261
PBS pH 7.2 (10mM NAH2PO4, 10mM Na2HPO4, 130mM NaCl) containing 0.1% bovine serum albumin.
−20°C. Avoid freeze-thaw cycles.
TRPC6