Learn More
Novus Biologicals™ VIP Overexpression Lysate (Native)
Supplier: Novus Biologicals™ NBL117722
Description
The VIP Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
0.1 mg | |
Protein | |
-80°C. Avoid freeze-thaw cycles. |
The lysate was created in HEK293T cells, using plasmid ID RC22452 and based on accession number NM_194435. The protein contains a C-terminal DDK tag. | |
RIPA buffer | |
Homo sapiens vasoactive intestinal peptide (VIP), transcript variant 2, mRNA. | |
Native |