Learn More
Novus Biologicals™ WFDC5 Overexpression Lysate (Native)
Supplier: Novus Biologicals™ NBL117842
Description
The WFDC5 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.Specifications
Western Blot | |
RIPA buffer | |
Homo sapiens WAP four-disulfide core domain 5 (WFDC5), mRNA. | |
Cancer, Cellular Signaling |
The lysate was created in HEK293T cells, using plasmid ID RC24523 and based on accession number NM_145652. The protein contains a C-terminal DDK tag. | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
Native |