Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Maleimide Activated R-Phycoerythrin

Applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen

Manufacturer:  Novus Biologicals™ 4030005

 View more versions of this product

Catalog No. 40-300-05

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The reactive maleimide group that has been added to the R-Phycoerythrin allows conjugation reactions to be performed very efficiently at physiological pH.

Dilutions
  • Immunohistochemistry 1:10-1:500
  • Immunohistochemistry- Frozen
  • Immunohistochemistry- Paraffin

Recommended Storage

Store at -20°C. Avoid freeze-thaw cycles.

Specifications

Specifications

5 mg
Maleimide Activated R-Phycoerythrin
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit