Learn More
Novus Biologicals™ Maleimide Activated R-Phycoerythrin
Applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
$760.00
Specifications
Product Type Maleimide Activated R-Phycoerythrin
View More Specs
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
40-300-02
View Documents
Novus Biologicals™
4030002
2mg
Each for $760.00
Add to cart
40-300-05
View Documents
Novus Biologicals™
4030005
5mg This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
The reactive maleimide group that has been added to the R-Phycoerythrin allows conjugation reactions to be performed very efficiently at physiological pH.
Dilutions
  • Immunohistochemistry 1:10-1:500
  • Immunohistochemistry- Frozen
  • Immunohistochemistry- Paraffin
Specifications
Specifications
Maleimide Activated R-Phycoerythrin