Learn More
Honeywell Burdick & Jackson™ NOWPak™ liner-based bottle system, Accessory, Pressure Dispense Connector (Bio)-NP516
For use with NOWPak™ Containers
Supplier:  Honeywell Burdick & Jackson™ NP516
Catalog No. 60-030-60
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Solvent can be dispensed at pressures less than 15 psig from NOWPak™ containers connected directly to DNA and peptide synthesizers. Each adapter consists of: A pressure dispense cap with PFA solvent fitting, an inert gas pressurization fitting and a 15 psig pressure relief valve. An FEP diptube with a 40-micron stainless steel frit to prevent drips while changing containers.
Specifications
Specifications
Each