Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nox4 Mouse anti-Human, Alexa Fluor™ 532, Clone: NOX4/1245, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP254338AF532
Description
Nox4 Monoclonal specifically detects Nox4 in Human samples. It is validated for Western Blot, ELISA.Specifications
Nox4 | |
NOX4/1245 | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
Purified, Conjugated | |
NOX4 | |
Recombinant human NOX4 protein | |
Protein A or G purified | |
Cancer, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Signal Transduction | |
Primary | |
50507.0 |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.- | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title