Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOX4 Mouse anti-Human, Clone: NOX4/1245, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14857

Catalog No. 89-034-940


Add to cart

Description

Description

Mouse monoclonal antibody raised against full length recombinant human NOX4.

  • High specificity
  • High reactivity
Specifications

Specifications

NOX4
NOX4/1245
Mouse monoclonal antibody raised against full length recombinant human NOX4.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Liquid
NOX4
Recombinant protein corresponding to full length human NOX4.
Protein A/G purification
RUO
Primary
50507
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q9NPH5
KOX|KOX-1|RENOX
Mouse
IgG2b, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)