Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nox4 Mouse anti-Human, Clone: NOX4/1245, Novus Biologicals™

Mouse Monoclonal Antibody

$175.29 - $410.59

Specifications

Antigen Nox4
Clone NOX4/1245
Host Species Mouse
Isotype IgG2b Kappa
Gene Alias EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.-
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25340320
View Documents
Novus Biologicals
NBP25340320UG
20 ug
Each for $175.29
Add to cart
 
NB253403100
View Documents
Novus Biologicals
NBP253403100UG
100 ug
Each for $410.59
Add to cart
 
NB253403200
View Documents
Novus Biologicals
NBP253403200UG
200 ug This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Nox4 Monoclonal specifically detects Nox4 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Nox4
Mouse
EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOXEC 1.6.3.-
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, SDS-Page
Unconjugated
Store at 4C.
Monoclonal
0.2 mg/mL
NOX4/1245
IgG2b Kappa
Human
RUO
Purified
Primary
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit