Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NOX5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543433
Description
Peptide sequence: DQAEEAQYGR FLELHMYMTS ALGKNDMKAI GLQMALDLLA NKEKKDSITG Sequence homology: Cow: 86%; Human: 100%; Pig: 79%; Rabbit: 100%.
This gene is predominantly expressed in the testis and lymphocyte-rich areas of spleen and lymph nodes. It encodes a calcium-dependen NADPH oxidase that generates superoxide, and functions as a calcium-dependent proton channel that may regulate redox-dependent processes in lymphocytes and spermatozoa. Alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Oct 2011].
Specifications
NOX5 | |
Polyclonal | |
Unconjugated | |
NOX5 | |
NADPH oxidase 5; NADPH oxidase, EF-hand calcium binding domain 5; NOX5; NOX5zeta | |
Rabbit | |
Affinity Chromatography | |
RUO | |
79400 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96PH1 | |
NOX5 | |
Synthetic peptide directed towards the C-terminal of human NOX5 (aa 659-708). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction