Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Noxa Rat anti-Mouse, Alexa Fluor™ 594, Clone: 69N15C9, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP225266AF594

Catalog No. NP22266A94


Add to cart

Description

Description

Noxa Monoclonal antibody specifically detects Noxa in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Noxa
69N15C9
Western Blot
adult T cell leukemia-derived PMA-responsive, APR, Immediate-early-response protein APR, NOXAPMA-induced protein 1, phorbol-12-myristate-13-acetate-induced protein 1, Protein Noxa
Rat
IgG2b Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
PMAIP1
Amino acids 1-103 of mouse Noxa were used as the immunogen for the antibody.
0.1 mL
Primary
5366
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit