Learn More
Abnova™ NOXA1 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00010811T01
Catalog No. 89-954-426
Add to cart
Description
Description
Specifications
Specifications
293T
NM_006647
pCMV-NOXA1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NOXA1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 51.7kDa
Transient overexpression cell lysate was tested with anti-NOXA1 antibody (Mfr. No. H00010811-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human