Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ NP-40 Alternative, Protein Grade™ Detergent, 10% Solution, Sterile-Filtered, Calbiochem™,

Click to view available options
Quantity:
50mL
500mL
Packaging:
Glass Bottle
9016-45-9
C19H32O3
308.462
BLXVTZPGEOGTGG-UHFFFAOYSA-N
Nonylphenyl Polyethylene Glycol
24773
2-[2-(4-nonylphenoxy)ethoxy]ethanol
CCCCCCCCCC1=CC=C(C=C1)OCCOCCO
9016-45-9
C19H32O3
308.462
BLXVTZPGEOGTGG-UHFFFAOYSA-N
Nonylphenyl Polyethylene Glycol
24773
2-[2-(4-nonylphenoxy)ethoxy]ethanol
CCCCCCCCCC1=CC=C(C=C1)OCCOCCO
Packaging Glass Bottle
Grade Protein
Color Undesignated
Quantity 500mL