Learn More
NP, Clone: B1-8, Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00192
Catalog No. 89-032-535
Add to cart
Specifications
Specifications
NP
B1-8
Flow Cytometry Negative Control Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2, lambda
RUO
Antibody
Monoclonal
Flow Cytometry, Western Blot
Unconjugated
In PBS (0.02% Proclin 300).
Original antibody is raised against hapten-protein conjugated NP12-chicken gammaglobulin (CG).
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
Hapten (4-hydroxy-3-nitropheny1) acetyl (NP).