Learn More
NP, Clone: B1-8, Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00166
Catalog No. 89-032-509
Add to cart
Specifications
Specifications
NP
B1-8
Flow Cytometry (1 ug/mL) Negative Control Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG2a, lambda
RUO
Antibody
Monoclonal
Flow Cytometry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against hapten-protein conjugated NP12-chicken gammaglobulin.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.