Learn More
Npas4 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S408-79, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259332FR
Catalog No. NBP259332FR
Add to cart
Description
Description
Npas4 Monoclonal antibody specifically detects Npas4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications
Specifications
Npas4
S408-79
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
NPAS4
Fusion protein amino acids 597-802 (C-terminus) of rat Npas4.
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 90kDa.
Western Blot, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
BHLHE79, bHLHe79neuronal PAS domain-containing protein 4, Class E basic helix-loop-helix protein 79, Le-PAS, neuronal PAS domain protein 4, Neuronal PAS4, NXFHLH-PAS transcription factor NXF, PASD10PAS domain-containing protein 10
Mouse
IgG1
100 ul
Primary
266743
Human, Mouse, Rat