Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Npas4 Mouse anti-Rat, PE/ATTO 594, Clone: S408-79, Abnova™

Mouse Monoclonal Antibody.

Manufacturer:  Abnova Corporation MAB19194

Catalog No. 89-338-698


Add to cart

Description

Description

Npas4 Monoclonal antibody specifically detects Npas4 in Rat samples. It is validated for ICC, WB-Ti, IHC-P, IF.
Specifications

Specifications

Npas4
S408-79
neuronal PAS domain protein 4
Liquid
Mouse
IgG1
Store at -20°C
Monoclonal
Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
PE/ATTO 594
Npas4
Nxf
Recombinant protein corresponding to amino acids 597-802 at C-terminus of rat Npas4
100 μg
Primary
266734
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit